missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcription activator BRG1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcription activator BRG1, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001122316.1 |
---|---|
Gene ID (Entrez) | 6597 |
Name | Transcription activator BRG1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028261
|
enQuireBio™
QP9406-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16068261
|
enQuireBio™
QP9406-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16038261
|
enQuireBio™
QP9406-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058261
|
enQuireBio™
QP9406-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16078261
|
enQuireBio™
QP9406-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048261
|
enQuireBio™
QP9406-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001122316.1 | |
Transcription activator BRG1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6597 | |
Research Use Only | |
SMARCA4 | |
Recombinant Protein | |
Yeast | |
EVDARHIIEN AKQDVDDEYG VSQALARGLQ SYYAVAHAVT ERVDKQSALM VNGVLKQYQI KGLEWLVSLY NNNLNGILAD EMGLGKTIQT IALITYLMEH KRINGPFLII VPLSTLSNWA YEFDKWAPSV VKVSYKGSPA ARRAFVPQLR SGKFNVLLTT YEYIIKDKHI LAKIRWKYMI VDEGHRMKNH HCKLTQVLNT HYVAPRRLLL TGTPLQNKLP ELWALLNFLL PTIFKSCSTF EQWFNAPFAM TGEKVDLNEE ETILIIRRLH KVLRPFLLRR LKKEVEAQLP EKVEYVIKCD MSALQRVLYR HMQAKGVLLT DGSEKDKKGK GGTKTLMNTI MQLRKICNHP YMFQHIEESF SEHLGFTGGI VQGLDLYRAS GKFELLDRIL PKLRATNHKV LLFCQMTSLM TIMEDYFAYR GFKYLRLDGT TKAEDRGMLL KTFNEPGSEY FIFLLSTRAG GLGLNLQSAD TVIIFDSDWN PHQDLQAQDR AHRIGQQNEV RVLRLCTVNS VEEKILAAAK YKLNVDQKVI QAGMFDQKSS SHERRAF | |
Greater than 90% as determined by SDS-PAGE. |