missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TPM4 / Tropomyosin 4 Protein
A DNA sequence encoding the Homo sapiens (Human) TPM4 / Tropomyosin 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6828-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138632.1 | |
TPM4 / Tropomyosin 4 Protein | |
Research Use Only | |
TPM4 | |
Recombinant Protein | |
E. coli | |
AGLNSLEAVK RKIQALQQQA DEAEDRAQGL QRELDGERER REKAEGDVAA LNRRIQLVEE ELDRAQERLA TALQKLEEAE KAADESERGM KVIENRAMKD EEKMEIQEMQ LKEAKHIAEE ADRKYEEVAR KLVILEGELE RAEERAEVSE LKCGDLEEEL KNVTNNLKSL EAASEKYSEK EDKYEEEIKL LSDKLKEAET RAEFAERTVA KLEKTIDDLE EKLAQAKEEN VGLHQTLDQT LNELNCI | |
Greater than 80% as determined by SDS-PAGE. |
7171 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |