missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thyroid peroxidase / TPO Protein
A DNA sequence encoding the Homo sapiens (Human) Thyroid peroxidase / TPO, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9441-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000538.3 | |
Thyroid peroxidase / TPO Protein | |
Research Use Only | |
TPO | |
Recombinant Protein | |
Yeast | |
FFPFISRGKE LLWGKPEESR VSSVLEESKR LVDTAMYATM QRNLKKRGIL SPAQLLSFSK LPEPTSGVIA RAAEIMETSI QAMKRKVNLK TQQSQHPTDA LSEDLLSIIA NMSGCLPYML PPKCPNTCLA NKYRPITGAC NNR | |
Greater than 90% as determined by SDS-PAGE. |
7173 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |