missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thyroid hormone receptor beta Protein

A DNA sequence encoding the Homo sapiens (Human) Thyroid hormone receptor beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6788-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015870

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_000452.2
Thyroid hormone receptor beta Protein
Research Use Only
THRB
Recombinant Protein
E. coli
MTPNSMTENG LTAWDKPKHC PDREHDWKLV GMSEACLHRK SHSERRSTLK NEQSSPHLIQ TTWTSSIFHL DHDDVNDQSV SSAQTFQTEE KKCKGYIPSY LDKDELCVVC GDKATGYHYR CITCEGCKGF FRRTIQKNLH PSYSCKYEGK CVIDKVTRNQ CQECRFKKCI YVGMATDLVL DDSKRLAKRK LIEENREKRR REELQKSIGH KPEPTDEEWE LIKTVTEAHV ATNAQGSHWK QKRKFLPEDI GQAPIVNAPE GGKVDLEAFS HFTKIITPAI TRVVDFAKKL PMFCELPCED QIILLKGCCM EIMSLRAAVR YDPESETLTL NGEMAVTRGQ LKNGGLGVVS DAIFDLGMSL SSFNLDDTEV ALLQAVLLMS SDRPGLACVE RIEKYQDSFL LAFEHYINYR KHHVTHFWPK LLMKVTDLRM IGACHASRFL HMKVECPTEL FPPLFLEVFE D
Greater than 90% as determined by SDS-PAGE.
7068
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers