missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thymidine kinase Protein
A DNA sequence encoding the Homo sapiens (Human) Thymidine kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6799-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
YP_009137097.1 | |
Thymidine kinase Protein | |
Research Use Only | |
TK | |
Recombinant Protein | |
E. coli | |
MASYPCHQHA SAFDQAARSR GHNNRRTALR PRRQQEATEV RPEQKMPTLL RVYIDGPHGM GKTTTTQLLV ALGSRDDIVY VPEPMTYWRV LGASETIANI YTTQHRLDQG EISAGDAAVV MTSAQITMGM PYAVTDAVLA PHIGGEAGSS HAPPPALTLI FDRHPIAALL CYPAARYLMG SMTPQAVLAF VALIPPTLPG TNIVLGALPE DRHIDRLAKR QRPGERLDLA MLAAIRRVYG LLANTVRYLQ CGGSWREDWG QLSGTAVPPQ GAEPQSNAGP RPHIGDTLFT LFRAPELLAP NGDLYNVFAW ALDVLAKRLR SMHVFILDYD QSPAGCRDAL LQLTSGMVQT HVTTPGSIPT ICDLARTFAR EMGEAN | |
Greater than 90% as determined by SDS-PAGE. |
24271467 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |