Learn More
enQuireBio™ Recombinant Human THAP domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) THAP domain-containing protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7886-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001304720.1 | |
THAP domain-containing protein 1 | |
Research Use Only | |
THAP6 | |
Recombinant Protein | |
E. coli | |
MVKCCSAIGC ASRCLPNSKL KGLTFHVFPT DENIKRKWVL AMKRLDVNAA GIWEPKKGDV LCSRHFKKTD FDRSAPNIKL KPGVIPSIFD SPYHLQGKRE KLHCRKNFTL KTVPATNYNH HLVGASSCIE EFQSQFIFEH SYSVMDSPKK LKHKLDHVIG ELEDTKESLR NVLDREKRFQ KSLRKTIREL KDECLISQET ANRLDTFCWD CCQESIEQDY IS | |
Greater than 90% as determined by SDS-PAGE. |
152815 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |