missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tegument protein VP16 Protein
A DNA sequence encoding the Homo sapiens (Human) Tegument protein VP16, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7282-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
Tegument protein VP16 Protein | |
Research Use Only | |
UL48 | |
Recombinant Protein | |
E. coli | |
MDLLVDELFA DMDADGASPP PPRPAGGPKN TPAAPPLYAT GRLSQAQLMP SPPMPVPPAA LFNRLLDDLG FSAGPALCTM LDTWNEDLFS ALPTNADLYR ECKFLSTLPS DVVEWGDAYV PERAQIDIRA HGDVAFPTLP ATRDGLGLYY EALSRFFHAE LRAREESYRT VLANFCSALY RYLRASVRQL HRQAHMRGRD RDLGEMLRAT IADRYYRETA RLARVLFLHL YLFLTREILW AAYAEQMMRP DLFDCLCCDL ESWRQLAGLF QPFMFVNGAL TVRGVPIEAR RLRELNHIRE HLNLPLVRSA ATEEPGAPLT TPPTLHGNQA RASGYFMVLI RAKLDSYSSF TTSPSEAVMR EHAYSRARTK NNYGSTIEGL LDLPDDDAPE EAGLAAPRLS FLPAGHTRRL STAPPTDVSL GDELHLDGED VAMAHADALD DFDLDMLGDG DSPGPGFTPH DSAPYGALDM ADFEFEQMFT DALGIDEYGG | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |