missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human T-box transcription factor TBX18 Protein

A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX18, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6766-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065680

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001073977.1
T-box transcription factor TBX18 Protein
Research Use Only
TBX18
Recombinant Protein
E. coli
MAEKRRGSPC SMLSLKAHAF SVEALIGAEK QQQLQKKRRK LGAEEAAGAV DDGGCSRGGG AGEKGSSEGD EGAALPPPAG ATSGPARSGA DLERGAAGGC EDGFQQGASP LASPGGSPKG SPARSLARPG TPLPSPQAPR VDLQGAELWK RFHEIGTEMI ITKAGRRMFP AMRVKISGLD PHQQYYIAMD IVPVDNKRYR YVYHSSKWMV AGNADSPVPP RVYIHPDSPA SGETWMRQVI SFDKLKLTNN ELDDQGHIIL HSMHKYQPRV HVIRKDCGDD LSPIKPVPSG EGVKAFSFPE TVFTTVTAYQ NQQITRLKID RNPFAKGFRD SGRNRMGLEA LVESYAFWRP SLRTLTFEDI PGIPKQGNAS SSTLLQGTGN GVPATHPHLL SGSSCSSPAF HLGPNTSQLC SLAPADYSAC ARSGLTLNRY STSLAETYNR LTNQAGETFA PPRTPSYVGV SSSTSVNMSM GGTDGDTFSC PQTSLSMQIS GMSPQLQYIM PSPSSNAFAT NQTHQGSYNT FRLHSPCALY GYNFSTSPKL AASPEKIVSS QGSFLGSSPS GTMTDRQMLP PVEGVHLLSS GGQQSFFDSR TLGSLTLSSS QVSAHMV
Greater than 90% as determined by SDS-PAGE.
9096
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers