missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human T-box transcription factor TBX18 Protein
A DNA sequence encoding the Homo sapiens (Human) T-box transcription factor TBX18, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6766-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001073977.1 | |
T-box transcription factor TBX18 Protein | |
Research Use Only | |
TBX18 | |
Recombinant Protein | |
E. coli | |
MAEKRRGSPC SMLSLKAHAF SVEALIGAEK QQQLQKKRRK LGAEEAAGAV DDGGCSRGGG AGEKGSSEGD EGAALPPPAG ATSGPARSGA DLERGAAGGC EDGFQQGASP LASPGGSPKG SPARSLARPG TPLPSPQAPR VDLQGAELWK RFHEIGTEMI ITKAGRRMFP AMRVKISGLD PHQQYYIAMD IVPVDNKRYR YVYHSSKWMV AGNADSPVPP RVYIHPDSPA SGETWMRQVI SFDKLKLTNN ELDDQGHIIL HSMHKYQPRV HVIRKDCGDD LSPIKPVPSG EGVKAFSFPE TVFTTVTAYQ NQQITRLKID RNPFAKGFRD SGRNRMGLEA LVESYAFWRP SLRTLTFEDI PGIPKQGNAS SSTLLQGTGN GVPATHPHLL SGSSCSSPAF HLGPNTSQLC SLAPADYSAC ARSGLTLNRY STSLAETYNR LTNQAGETFA PPRTPSYVGV SSSTSVNMSM GGTDGDTFSC PQTSLSMQIS GMSPQLQYIM PSPSSNAFAT NQTHQGSYNT FRLHSPCALY GYNFSTSPKL AASPEKIVSS QGSFLGSSPS GTMTDRQMLP PVEGVHLLSS GGQQSFFDSR TLGSLTLSSS QVSAHMV | |
Greater than 90% as determined by SDS-PAGE. |
9096 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |