missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Syndecan-4 / SDC4 Protein

A DNA sequence encoding the Homo sapiens (Human) Syndecan-4 / SDC4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7123-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047440

  • 515.00€ / 50µg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_002990.2
Syndecan-4 / SDC4 Protein
Research Use Only
SDC4
Recombinant Protein
E. coli
ESIRETEVID PQDLLEGRYF SGALPDDEDV VGPGQESDDF ELSGSGDLDD LEDSMIGPEV VHPLVPLDNH IPERAGSGSQ VPTEPKKLEE NEVIPKRISP VEESEDVSNK VSMSSTVQGS NIFERTE
Greater than 90% as determined by SDS-PAGE.
6385
50 μg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers