missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SULT1A1 / STP1 Protein
A DNA sequence encoding the Homo sapiens (Human) SULT1A1 / STP1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6746-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001046.2 | |
SULT1A1 / STP1 Protein | |
Research Use Only | |
SULT1A1 | |
Recombinant Protein | |
E. coli | |
MELIQDTSRP PLEYVKGVPL IKYFAEALGP LQSFQARPDD LLISTYPKSG TTWVSQILDM IYQGGDLEKC HRAPIFMRVP FLEFKAPGIP SGMETLKDTP APRLLKTHLP LALLPQTLLD QKVKVVYVAR NAKDVAVSYY HFYHMAKVHP EPGTWDSFLE KFMVGEVSYG SWYQHVQEWW ELSRTHPVLY LFYEDMKENP KREIQKILEF VGHSLPEETV DFVVQHTSFK EMKKNPMTNY TTVPQEFMDH SISPFMRKGM AGDWKTTFTV AQNERFDADY AEKMAGCSLS FRSEL | |
Greater than 80% as determined by SDS-PAGE. |
6817 | |
10 μg | |
Not determined. | |
Full Length of BC000923 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |