missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sulfite oxidase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Sulfite oxidase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6750-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015570

  • 1740.00€ / 500µg
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_000447.2
Sulfite oxidase, mitochondrial Protein
Research Use Only
SUOX
Recombinant Protein
Yeast
ESTHIYTKEE VSSHTSPETG IWVTLGSEVF DVTEFVDLHP GGPSKLMLAA GGPLEPFWAL YAVHNQSHVR ELLAQYKIGE LNPEDKVAPT VETSDPYADD PVRHPALKVN SQRPFNAEPP PELLTENYIT PNPIFFTRNH LPVPNLDPDT YRLHVVGAPG GQSLSLSLDD LHNFPRYEIT VTLQCAGNRR SEMTQVKEVK GLEWRTGAIS TARWAGARLC DVLAQAGHQL CETEAHVCFE GLDSDPTGTA YGASIPLARA MDPEAEVLLA YEMNGQPLPR DHGFPVRVVV PGVVGARHVK WLGRVSVQPE ESYSHWQRRD YKGFSPSVDW ETVDFDSAPS IQELPVQSAI TEPRDGETVE SGEVTIKGYA WSGGGRAVIR VDVSLDGGLT WQVAKLDGEE QRPRKAWAWR LWQLKAPVPA GQKELNIVCK AVDDGYNVQP DTVAPIWNLR GVLSNAWHRV HVYVSP
Greater than 90% as determined by SDS-PAGE.
6821
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers