missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sulfite oxidase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Sulfite oxidase, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6750-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000447.2 | |
Sulfite oxidase, mitochondrial Protein | |
Research Use Only | |
SUOX | |
Recombinant Protein | |
Yeast | |
ESTHIYTKEE VSSHTSPETG IWVTLGSEVF DVTEFVDLHP GGPSKLMLAA GGPLEPFWAL YAVHNQSHVR ELLAQYKIGE LNPEDKVAPT VETSDPYADD PVRHPALKVN SQRPFNAEPP PELLTENYIT PNPIFFTRNH LPVPNLDPDT YRLHVVGAPG GQSLSLSLDD LHNFPRYEIT VTLQCAGNRR SEMTQVKEVK GLEWRTGAIS TARWAGARLC DVLAQAGHQL CETEAHVCFE GLDSDPTGTA YGASIPLARA MDPEAEVLLA YEMNGQPLPR DHGFPVRVVV PGVVGARHVK WLGRVSVQPE ESYSHWQRRD YKGFSPSVDW ETVDFDSAPS IQELPVQSAI TEPRDGETVE SGEVTIKGYA WSGGGRAVIR VDVSLDGGLT WQVAKLDGEE QRPRKAWAWR LWQLKAPVPA GQKELNIVCK AVDDGYNVQP DTVAPIWNLR GVLSNAWHRV HVYVSP | |
Greater than 90% as determined by SDS-PAGE. |
6821 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |