missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STAT3 Protein

A DNA sequence encoding the Homo sapiens (Human) STAT3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6735-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16045470

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_003141.2
STAT3 Protein
Research Use Only
STAT3
Recombinant Protein
E. coli
ESHATLVFHN LLGEIDQQYS RFLQESNVLY QHNLRRIKQF LQSRYLEKPM EIARIVARCL WEESRLLQTA ATAAQQGGQA NHPTAAVVTE KQQMLEQHLQ DVRKRVQDLE QKMKVVENLQ DDFDFNYKTL KSQGDMQDLN GNNQSVTRQK MQQLEQMLTA LDQMRRSIVS ELAGLLSAME YVQKTLTDEE L
Greater than 90% as determined by SDS-PAGE.
6774
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers