missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Spindlin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Spindlin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8292-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013821

  • 530.00€ / 100µg
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006708.2
Spindlin-1 Protein
Research Use Only
SPIN1
Recombinant Protein
E. coli
MKTPFGKTPG QRSRADAGHA GVSANMMKKR TSHKKHRSSV GPSKPVSQPR RNIVGCRIQH GWKEGNGPVT QWKGTVLDQV PVNPSLYLIK YDGFDCVYGL ELNKDERVSA LEVLPDRVAT SRISDAHLAD TMIGKAVEHM FETEDGSKDE WRGMVLARAP VMNTWFYITY EKDPVLYMYQ LLDDYKEGDL RIMPDSNDSP PAEREPGEVV DSLVGKQVEY AKEDGSKRTG MVIHQVEAKP SVYFIKFDDD FHIYVYDLVK TS
Greater than 80% as determined by SDS-PAGE.
10927
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers