missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SOSS complex subunit B2 Protein

A DNA sequence encoding the Homo sapiens (Human) SOSS complex subunit B2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7959-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061841

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001026886.1
SOSS complex subunit B2 Protein
Research Use Only
NABP1
Recombinant Protein
E. coli
MNRVNDPLIF IRDIKPGLKN LNVVFIVLEI GRVTKTKDGH EVRSCKVADK TGSITISVWD EIGGLIQPGD IIRLTRGYAS MWKGCLTLYT GRGGELQKIG EFCMVYSEVP NFSEPNPDYR GQQNKGAQSE QKNNSMNSNM GTGTFGPVGN GVHTGPESRE HQFSHAGRSN GRGLINPQLQ GTASNQTVMT TISNGRDPRR AFKR
Greater than 90% as determined by SDS-PAGE.
64859
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers