missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SOSS complex subunit B2 Protein
A DNA sequence encoding the Homo sapiens (Human) SOSS complex subunit B2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7959-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001026886.1 | |
SOSS complex subunit B2 Protein | |
Research Use Only | |
NABP1 | |
Recombinant Protein | |
E. coli | |
MNRVNDPLIF IRDIKPGLKN LNVVFIVLEI GRVTKTKDGH EVRSCKVADK TGSITISVWD EIGGLIQPGD IIRLTRGYAS MWKGCLTLYT GRGGELQKIG EFCMVYSEVP NFSEPNPDYR GQQNKGAQSE QKNNSMNSNM GTGTFGPVGN GVHTGPESRE HQFSHAGRSN GRGLINPQLQ GTASNQTVMT TISNGRDPRR AFKR | |
Greater than 90% as determined by SDS-PAGE. |
64859 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |