missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sorting nexin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7609-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001229862.1 | |
Sorting nexin-1 Protein | |
Research Use Only | |
SNX1 | |
Recombinant Protein | |
E. coli | |
MASGGGGCSA SERLPPPFPG LEPESEGAAG GSEPEAGDSD TEGEDIFTGA AVVSKHQSPK ITTSLLPINN GSKENGIHEE QDQEPQDLFA DATVELSLDS TQNNQKKVLA KTLISLPPQE ATNSSKPQPT YEELEEEEQE DQFDLTVGIT DPEKIGDGMN AYVAYKVTTQ TSLPLFRSKQ FAVKRRFSDF LGLYEKLSEK HSQNGFIVPP PPEKSLIGMT KVKVGKEDSS SAEFLEKRRA ALERYLQRIV NHPTMLQDPD VREFLEKEEL PRAVGTQTLS GAGLLKMFNK ATDAVSKMTI KMNESDIWFE EKLQEVECEE QRLRKLHAVV ETLVNHRKEL ALNTAQFAKS LAMLGSSEDN TALSRALSQL AEVEEKIEQL HQEQANNDFF LLAELLSDYI RLLAIVRAAF DQRMKTWQRW QDAQATLQKK REAEARLLWA NKPDKLQQAK DEILEWESRV TQYERDFERI STVVRKEVIR FEKEKSKDFK NHVIKYLETL LYSQQQLAKY WEAFLPEAKA IS | |
Greater than 90% as determined by SDS-PAGE. |
6642 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |