missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sorting nexin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7609-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039750

  • 2385.00€ / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001229862.1
Sorting nexin-1 Protein
Research Use Only
SNX1
Recombinant Protein
E. coli
MASGGGGCSA SERLPPPFPG LEPESEGAAG GSEPEAGDSD TEGEDIFTGA AVVSKHQSPK ITTSLLPINN GSKENGIHEE QDQEPQDLFA DATVELSLDS TQNNQKKVLA KTLISLPPQE ATNSSKPQPT YEELEEEEQE DQFDLTVGIT DPEKIGDGMN AYVAYKVTTQ TSLPLFRSKQ FAVKRRFSDF LGLYEKLSEK HSQNGFIVPP PPEKSLIGMT KVKVGKEDSS SAEFLEKRRA ALERYLQRIV NHPTMLQDPD VREFLEKEEL PRAVGTQTLS GAGLLKMFNK ATDAVSKMTI KMNESDIWFE EKLQEVECEE QRLRKLHAVV ETLVNHRKEL ALNTAQFAKS LAMLGSSEDN TALSRALSQL AEVEEKIEQL HQEQANNDFF LLAELLSDYI RLLAIVRAAF DQRMKTWQRW QDAQATLQKK REAEARLLWA NKPDKLQQAK DEILEWESRV TQYERDFERI STVVRKEVIR FEKEKSKDFK NHVIKYLETL LYSQQQLAKY WEAFLPEAKA IS
Greater than 90% as determined by SDS-PAGE.
6642
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers