missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SMARCA2 Protein
A DNA sequence encoding the Homo sapiens (Human) SMARCA2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6698-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001276325.1 | |
SMARCA2 Protein | |
Research Use Only | |
SMARCA2 | |
Recombinant Protein | |
Yeast | |
SYYTVAHAIS ERVEKQSALL INGTLKHYQL QGLEWMVSLY NNNLNGILAD EMGLGKTIQT IALITYLMEH KRLNGPYLII VPLSTLSNWT YEFDKWAPSV VKISYKGTPA MRRSLVPQLR SGKFNVLLTT YEYIIKDKHI LAKIRWKYMI VDEGHRMKNH HCKLTQVLNT HYVAPRRILL TGTPLQNKLP ELWALLNFLL PTIFKSCSTF EQWFNAPFAM TGERVDLNEE ETILIIRRLH KVLRPFLLRR LKKEVESQLP EKVEYVIKCD MSALQKILYR HMQAKGILLT DGSEKDKKGK GGAKTLMNTI MQLRKICNHP YMFQHIEESF AEHLGYSNGV INGAELYRAS GKFELLDRIL PKLRATNHRV LLFCQMTSLM TIMEDYFAFR NFLYLRLDGT TKSEDRAALL KKFNEPGSQY FIFLLSTRAG GLGLNLQAAD TVVIFDSDWN PHQDLQAQDR AHRIGQQNEV RVLRLCTVNS VEEKILAAAK YKLNVDQKVI QAGMFDQKSS SHERRAF | |
Greater than 90% as determined by SDS-PAGE. |
6595 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |