missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small proline-rich protein 1

A DNA sequence encoding the Homo sapiens (Human) Small proline-rich protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6724-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015400

  • 2385.00€ / 1mg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001091058.1
Small proline-rich protein 1
Research Use Only
SPRR3
Recombinant Protein
E. coli
SSYQQKQTFT PPPQLQQQQV KQPSQPPPQE IFVPTTKEPC HSKVPQPGNT KIPEPGCTKV PEPGCTKVPE PGCTKVPEPG CTKVPEPGCT KVPEPGCTKV PEPGYTKVPE PGSIKVPDQG FIKFPEPGAI KVPEQGYTKV PVPGYTKLPE PCPSTVTPGP AQQKTKQK
Greater than 90% as determined by SDS-PAGE.
6707
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers