missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Small proline-rich protein 1
A DNA sequence encoding the Homo sapiens (Human) Small proline-rich protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6724-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001091058.1 | |
Small proline-rich protein 1 | |
Research Use Only | |
SPRR3 | |
Recombinant Protein | |
E. coli | |
SSYQQKQTFT PPPQLQQQQV KQPSQPPPQE IFVPTTKEPC HSKVPQPGNT KIPEPGCTKV PEPGCTKVPE PGCTKVPEPG CTKVPEPGCT KVPEPGCTKV PEPGYTKVPE PGSIKVPDQG FIKFPEPGAI KVPEQGYTKV PVPGYTKLPE PCPSTVTPGP AQQKTKQK | |
Greater than 90% as determined by SDS-PAGE. |
6707 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |