missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small proline-rich protein 1

A DNA sequence encoding the Homo sapiens (Human) Small proline-rich protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6724-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16035400

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001091058.1
Small proline-rich protein 1
Research Use Only
SPRR3
Recombinant Protein
E. coli
SSYQQKQTFT PPPQLQQQQV KQPSQPPPQE IFVPTTKEPC HSKVPQPGNT KIPEPGCTKV PEPGCTKVPE PGCTKVPEPG CTKVPEPGCT KVPEPGCTKV PEPGYTKVPE PGSIKVPDQG FIKFPEPGAI KVPEQGYTKV PVPGYTKLPE PCPSTVTPGP AQQKTKQK
Greater than 90% as determined by SDS-PAGE.
6707
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers