missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Signal-regulatory protein beta-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Signal-regulatory protein beta-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6685-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001077379.1 | |
Signal-regulatory protein beta-1 Protein | |
Research Use Only | |
SIRPB1 | |
Recombinant Protein | |
Yeast | |
EDELQVIQPE KSVSVAAGES ATLRCAMTSL IPVGPIMWFR GAGAGRELIY NQKEGHFPRV TTVSELTKRN NLDFSISISN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAVRA TPEHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP AGDSVSYSIH STARVVLTRG DVHSQVICEI AHITLQGDPL RGTANLSEAI RVPPTLEVTQ QPMRAENQAN VTCQVSNFYP RGLQLTWLEN GNVSRTETAS TLIENKDGTY NWMSWLLVNT CAHRDDVVLT CQVEHDGQQA VSKSYALEIS AHQKEHGSDI THEAALAPTA PL | |
Greater than 90% as determined by SDS-PAGE. |
10326 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |