missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Signal recognition particle 14 kDa Protein

A DNA sequence encoding the Homo sapiens (Human) Signal recognition particle 14 kDa protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8428-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054561

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_003125.3
Signal recognition particle 14 kDa Protein
Research Use Only
SRP14
Recombinant Protein
E. coli
VLLESEQFLT ELTRLFQKCR TSGSVYITLK KYDGRTKPIP KKGTVEGFEP ADNKCLLRAT DGKKKISTVV SSKEVNKFQM AYSNLLRANM DGLKKRDKKN KTKKTKA
Greater than 90% as determined by SDS-PAGE.
6727
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers