missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / threonine-protein kinase STK11 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase STK11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7621-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000446.1 | |
Serine / threonine-protein kinase STK11 Protein | |
Research Use Only | |
STK11 | |
Recombinant Protein | |
E. coli | |
MEVVDPQQLG MFTEGELMSV GMDTFIHRID STEVIYQPRR KRAKLIGKYL MGDLLGEGSY GKVKEVLDSE TLCRRAVKIL KKKKLRRIPN GEANVKKEIQ LLRRLRHKNV IQLVDVLYNE EKQKMYMVME YCVCGMQEML DSVPEKRFPV CQAHGYFCQL IDGLEYLHSQ GIVHKDIKPG NLLLTTGGTL KISDLGVAEA LHPFAADDTC RTSQGSPAFQ PPEIANGLDT FSGFKVDIWS AGVTLYNITT GLYPFEGDNI YKLFENIGKG SYAIPGDCGP PLSDLLKGML EYEPAKRFSI RQIRQHSWFR KKHPPAEAPV PIPPSPDTKD RWRSMTVVPY LEDLHGADED EDLFDIEDDI IYTQDFTVPG QVPEEEASHN GQRRGLPKAV CMNGTEAAQL STKSRAEGRA PNPARKACSA SSKIRRLSAC | |
Greater than 90% as determined by SDS-PAGE. |
6794 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |