missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine / threonine-protein kinase STK11 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase STK11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7621-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029850

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_000446.1
Serine / threonine-protein kinase STK11 Protein
Research Use Only
STK11
Recombinant Protein
E. coli
MEVVDPQQLG MFTEGELMSV GMDTFIHRID STEVIYQPRR KRAKLIGKYL MGDLLGEGSY GKVKEVLDSE TLCRRAVKIL KKKKLRRIPN GEANVKKEIQ LLRRLRHKNV IQLVDVLYNE EKQKMYMVME YCVCGMQEML DSVPEKRFPV CQAHGYFCQL IDGLEYLHSQ GIVHKDIKPG NLLLTTGGTL KISDLGVAEA LHPFAADDTC RTSQGSPAFQ PPEIANGLDT FSGFKVDIWS AGVTLYNITT GLYPFEGDNI YKLFENIGKG SYAIPGDCGP PLSDLLKGML EYEPAKRFSI RQIRQHSWFR KKHPPAEAPV PIPPSPDTKD RWRSMTVVPY LEDLHGADED EDLFDIEDDI IYTQDFTVPG QVPEEEASHN GQRRGLPKAV CMNGTEAAQL STKSRAEGRA PNPARKACSA SSKIRRLSAC
Greater than 90% as determined by SDS-PAGE.
6794
50 μg
Not determined.
Full Length of Mature Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers