missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / threonine-protein kinase PAK 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase PAK 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7527-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001122092.1 | |
Serine / threonine-protein kinase PAK 1 Protein | |
Research Use Only | |
PAK1 | |
Recombinant Protein | |
E. coli | |
MSNNGLDIQD KPPAPPMRNT STMIGAGSKD AGTLNHGSKP LPPNPEEKKK KDRFYRSILP GDKTNKKKEK ERPEISLPSD FEHTIHVGFD AVTGEFTGMP EQWARLLQTS NITKSEQKKN PQAVLDVLEF YNSKKTSNSQ KYMSFTDKSA EDYNSSNALN VKAVSETPAV PPVSEDEDDD DDDATPPPVI APRPEHTKSV YTRSVIEPLP VTPTRDVATS PISPTENNTT PPDALTRNTE KQKKKPKMSD EEILEKLRSI VSVGDPKKKY TRFEKIGQGA SGTVYTAMDV ATGQEVAIKQ MNLQQQPKKE LIINEILVMR ENKNPNIVNY LDSYLVGDEL WVVMEYLAGG SLTDVVTETC MDEGQIAAVC RECLQALEFL HSNQVIHRDI KSDNILLGMD GSVKLTDFGF CAQITPEQSK RSTMVGTPYW MAPEVVTRKA YGPKVDIWSL GIMAIEMIEG EPPYLNENPL RALYLIATNG TPELQNPEKL SAIFRDFLNR CLEMDVEKRG SAKELLQHQF LKIAKPLSSL TPLIAAAKEA TKNNH | |
Greater than 90% as determined by SDS-PAGE. |
5058 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |