missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Septin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Septin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7552-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059340

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001008491.1
Septin-2 Protein
Research Use Only
42980
Recombinant Protein
E. coli
MSKQQPTQFI NPETPGYVGF ANLPNQVHRK SVKKGFEFTL MVVGESGLGK STLINSLFLT DLYPERVIPG AAEKIERTVQ IEASTVEIEE RGVKLRLTVV DTPGYGDAIN CRDCFKTIIS YIDEQFERYL HDESGLNRRH IIDNRVHCCF YFISPFGHGL KPLDVAFMKA IHNKVNIVPV IAKADTLTLK ERERLKKRIL DEIEEHNIKI YHLPDAESDE DEDFKEQTRL LKASIPFSVV GSNQLIEAKG KKVRGRLYPW GVVEVENPEH NDFLKLRTML ITHMQDLQEV TQDLHYENFR SERLKRGGRK VENEDMNKDQ ILLEKEAELR RMQEMIARMQ AQMQMQMQGG DGDGGALGHH V
Greater than 90% as determined by SDS-PAGE.
4735
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers