missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Secreted Ly-6 / uPAR-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Secreted Ly-6/uPAR-related protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6696-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_065160.1 | |
Secreted Ly-6 / uPAR-related protein 1 | |
Research Use Only | |
SLURP1 | |
Recombinant Protein | |
E. coli | |
LKCYTCKEPM TSASCRTITR CKPEDTACMT TLVTVEAEYP FNQSPVVTRS CSSSCVATDP DSIGAAHLIF CCFRDLCNSE L | |
Greater than 90% as determined by SDS-PAGE. |
57152 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |