missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RRM1 Protein

A DNA sequence encoding the Homo sapiens (Human) RRM1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6629-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044670

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001024.1
RRM1 Protein
Research Use Only
RRM1
Recombinant Protein
Yeast
MHVIKRDGRQ ERVMFDKITS RIQKLCYGLN MDFVDPAQIT MKVIQGLYSG VTTVELDTLA AETAATLTTK HPDYAILAAR IAVSNLHKET KKVFSDVMED LYNYINPHNG KHSPMVAKST LDIVLANKDR LNSAIIYDRD FSYNYFGFKT LERSYLLKIN GKVAERPQHM LMRVSVGIHK EDIDAAIETY NLLSERWFTH ASPTLFNAGT NRPQLSSCFL LSMKDDSIEG IYDTLKQCAL ISKSAGGIGV AVSCIRATGS YIAGTNGNSN GLVPMLRVYN NTARYVDQGG NKRPGAFAIY LEPWHLDIFE FLDLKKNTGK EEQRARDLFF ALWIPDLFMK RVETNQDWSL MCPNECPGLD EVWGEEFEKL YASYEKQGRV RKVVKAQQLW YAIIESQTET GTPYMLYKDS CNRKSNQQNL GTIKCSNLCT EIVEYTSKDE VAVCNLASLA LNMYVTSEHT YDFKKLAEVT KVVVRNLNKI IDINYYPVPE ACLSNKRHRP IGIGVQGLAD AFILMRYPFE SAEAQLLNKQ IFETIYYGAL EASCDLAKEQ GPYETYEGSP VSKGILQYDM WNVTPTDLWD WKVLKEKIAK YGIRNSLLIA PMPTASTAQI LGNNESIEPY TSNIYTRRVL SGEFQIVNPH LLKDLTERGL WHEEMKNQII ACNGSIQSIP EIPDDLKQLY KTVWEISQKT VLKMAAERGA FIDQSQSLNI HIAEPNYGKL TSMHFYGWKQ GLKTGMYYLR TRPAANPIQF TLNKEKLKDK EKVSKEEEEK ERNTAAMVCS LENRDECLMC GS
Greater than 90% as determined by SDS-PAGE.
6240
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers