missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNA-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) RNA-binding protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6586-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006734.1 | |
RNA-binding protein 1 | |
Research Use Only | |
RBM3 | |
Recombinant Protein | |
Yeast | |
MSSEEGKLFV GGLNFNTDEQ ALEDHFSSFG PISEVVVVKD RETQRSRGFG FITFTNPEHA SVAMRAMNGE SLDGRQIRVD HAGKSARGTR GGGFGAHGRG RSYSRGGGDQ GYGSGRYYDS RPGGYGYGYG RSRDYNGRNQ GGYDRYSGGN YRDNYDN | |
Greater than 90% as determined by SDS-PAGE. |
5935 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |