missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Retnlb Protein

A DNA sequence encoding the Homo sapiens (Human) Retnlb, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8034-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032281

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_115968.1
Retnlb Protein
Research Use Only
RETNLB
Recombinant Protein
E. coli
QCSLDSVMDK KIKDVLNSLE YSPSPISKKL SCASVKSQGR PSSCPAGMAV TGCACGYGCG SWDVQLETTC HCQCSVVDWT TARCCHLT
Greater than 90% as determined by SDS-PAGE.
84666
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers