missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RELT / TNFRSF19L Protein
A DNA sequence encoding the Homo sapiens (Human) RELT / TNFRSF19L, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8596-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116260.2 | |
RELT / TNFRSF19L Protein | |
Research Use Only | |
RELT | |
Recombinant Protein | |
E. coli | |
STTLWQCPPG EEPDLDPGQG TLCRPCPPGT FSAAWGSSPC QPHARCSLWR RLEAQVGMAT RDTLCGDCWP GWFGPWGVPR VPCQPCSWAP LGTHGCDEWG RRARRGVEVA AGASSGGETR QPGNGTRA | |
Greater than 90% as determined by SDS-PAGE. |
84957 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |