missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RELT / TNFRSF19L Protein

A DNA sequence encoding the Homo sapiens (Human) RELT / TNFRSF19L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8596-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005491

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_116260.2
RELT / TNFRSF19L Protein
Research Use Only
RELT
Recombinant Protein
E. coli
STTLWQCPPG EEPDLDPGQG TLCRPCPPGT FSAAWGSSPC QPHARCSLWR RLEAQVGMAT RDTLCGDCWP GWFGPWGVPR VPCQPCSWAP LGTHGCDEWG RRARRGVEVA AGASSGGETR QPGNGTRA
Greater than 90% as determined by SDS-PAGE.
84957
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers