missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulatory protein E2 Protein
A DNA sequence encoding the Homo sapiens (Human) Regulatory protein E2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7408-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_041328.1 | |
Regulatory protein E2 Protein | |
Research Use Only | |
E2 | |
Recombinant Protein | |
E. coli | |
METLCQRLNV CQDKILTHYE NDSTDLRDHI DYWKHMRLEC AIYYKAREMG FKHINHQVVP TLAVSKNKAL QAIELQLTLE TIYNSQYSNE KWTLQDVSLE VYLTAPTGCI KKHGYTVEVQ FDGDICNTMH YTNWTHIYIC EEASVTVVEG QVDYYGLYYV HEGIRTYFVQ FKDDAEKYSK NKVWEVHAGG QVILCPTSVF SSNEVSSPEI IRQHLANHPA ATHTKAVALG TEETQTTIQR PRSEPDTGNP CHTTKLLHRD SVDSAPILTA FNSSHKGRIN CNSNTTPIVH LKGDANTLKC LRYRFKKHCT LYTAVSSTWH WTGHNVKHKS AIVTLTYDSE WQRDQFLSQV KIPKTITVST GFMSI | |
Greater than 90% as determined by SDS-PAGE. |
1489080 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |