missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Recoverin Protein
A DNA sequence encoding the Homo sapiens (Human) Recoverin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6591-ye-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002894.1 | |
Recoverin Protein | |
Research Use Only | |
RCVRN | |
Recombinant Protein | |
Yeast | |
GNSKSGALSK EILEELQLNT KFSEEELCSW YQSFLKDCPT GRITQQQFQS IYAKFFPDTD PKAYAQHVFR SFDSNLDGTL DFKEYVIALH MTTAGKTNQK LEWAFSLYDV DGNGTISKNE VLEIVMAIFK MITPEDVKLL PDDENTPEKR AEKIWKYFGK NDDDKLTEKE FIEGTLANKE ILRLIQFEPQ KVKEKMKNA | |
Greater than 90% as determined by SDS-PAGE. |
5957 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |