missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ras-related protein Rab-17 Protein

A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-17, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8065-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072461

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_071894.1
Ras-related protein Rab-17 Protein
Research Use Only
RAB17
Recombinant Protein
E. coli
MAQAHRTPQP RAAPSQPRVF KLVLLGSGSV GKSSLALRYV KNDFKSILPT VGCAFFTKVV DVGATSLKLE IWDTAGQEKY HSVCHLYFRG ANAALLVYDI TRKDSFLKAQ QWLKDLEEEL HPGEVLVMLV GNKTDLSQER EVTFQEGKEF ADSQKLLFME TSAKLNHQVS EVFNTVAQEL LQRSDEEGQA LRGDAAVALN KGPARQAKCC AH
Greater than 80% as determined by SDS-PAGE.
64284
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers