missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-17 Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-17, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8065-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_071894.1 | |
Ras-related protein Rab-17 Protein | |
Research Use Only | |
RAB17 | |
Recombinant Protein | |
E. coli | |
MAQAHRTPQP RAAPSQPRVF KLVLLGSGSV GKSSLALRYV KNDFKSILPT VGCAFFTKVV DVGATSLKLE IWDTAGQEKY HSVCHLYFRG ANAALLVYDI TRKDSFLKAQ QWLKDLEEEL HPGEVLVMLV GNKTDLSQER EVTFQEGKEF ADSQKLLFME TSAKLNHQVS EVFNTVAQEL LQRSDEEGQA LRGDAAVALN KGPARQAKCC AH | |
Greater than 80% as determined by SDS-PAGE. |
64284 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |