missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-11A Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-11A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6566-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001193765.1 | |
Ras-related protein Rab-11A Protein | |
Research Use Only | |
RAB11A | |
Recombinant Protein | |
E. coli | |
GTRDDEYDYL FKVVLIGDSG VGKSNLLSRF TRNEFNLESK STIGVEFATR SIQVDGKTIK AQIWDTAGQE RYRAITSAYY RGAVGALLVY DIAKHLTYEN VERWLKELRD HADSNIVIML VGNKSDLRHL RAVPTDEARA FAEKNGLSFI ETSALDSTNV EAAFQTILTE IYRIVSQKQM SDRRENDMSP SNNVVPIHVP PTTENKPKVQ CC | |
Greater than 90% as determined by SDS-PAGE. |
8766 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |