missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PYCR1 Protein

A DNA sequence encoding the Homo sapiens (Human) PYCR1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6560-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064160

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_008838.2
PYCR1 Protein
Research Use Only
PYCR1
Recombinant Protein
E. coli
MEQLLSSVGF CTEVEEDLID AVTGLSGSGP AYAFTALDAL ADGGVKMGLP RRLAVRLGAQ ALLGAAKMLL HSEQHPGQLK DNVSSPGGAT IHALHVLESG GFRSLLINAV EASCIRTREL QSMADQEQVS PAAIKKTILD KVKLDSPAGT ALSPSGHTKL LPRSLAPAGK D
Greater than 90% as determined by SDS-PAGE.
5831
200 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers