missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PYCR1 Protein
A DNA sequence encoding the Homo sapiens (Human) PYCR1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6560-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_008838.2 | |
PYCR1 Protein | |
Research Use Only | |
PYCR1 | |
Recombinant Protein | |
E. coli | |
MEQLLSSVGF CTEVEEDLID AVTGLSGSGP AYAFTALDAL ADGGVKMGLP RRLAVRLGAQ ALLGAAKMLL HSEQHPGQLK DNVSSPGGAT IHALHVLESG GFRSLLINAV EASCIRTREL QSMADQEQVS PAAIKKTILD KVKLDSPAGT ALSPSGHTKL LPRSLAPAGK D | |
Greater than 90% as determined by SDS-PAGE. |
5831 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |