missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTPRZ1 Protein

A DNA sequence encoding the Homo sapiens (Human) PTPRZ1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6557-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16014140

  • 2385.00€ / 1mg
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001193767.1
PTPRZ1 Protein
Research Use Only
PTPRZ1
Recombinant Protein
E. coli
IGWSYTGALN QKNWGKKYPT CNSPKQSPIN IDEDLTQVNV NLKKLKFQGW DKTSLENTFI HNTGKTVEIN LTNDYRVSGG VSEMVFKASK ITFHWGKCNM SSDGSEHSLE GQKFPLEMQI YCFDADRFSS FEEAVKGKGK LRALSILFEV GTEENLDFKA IIDGVESVSR FGKQAALDPF ILLNLLPNST DKYYIYNGSL TSPPCTDTVD WIVFKDTVSI SESQLAVFCE VLTMQQSGYV MLMDYLQNNF REQQYKFSRQ VFSSY
Greater than 90% as determined by SDS-PAGE.
5803
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers