missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPN6 Protein
A DNA sequence encoding the Homo sapiens (Human) PTPN6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6555-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002822.2 | |
PTPN6 Protein | |
Research Use Only | |
PTPN6 | |
Recombinant Protein | |
E. coli | |
MLSRGVGDQV THIRIQNSGD FYDLYGGEKF ATLTELVEYY TQQQGVLQDR DGTIIHLKYP LNCSDPTSER WYHGHMSGGQ AETLLQAKGE PWTFLVRESL SQPGDFVLSV LSDQPKAGPG SPLRVTHIKV MCEGGRYTVG GLETFDSLTD LVEHFKKTGI EEASGAFVYL RQPYYATRVN AADIENRVLE LNKKQESEDT AKAGFWEEFE SLQKQEVKNL HQRLEGQRPE NKGKNRYKNI LPFDHSRVIL QGRDSNIPGS DYINANYIKN QLLGPDENAK TYIASQGCLE ATVNDFWQMA WQENSRVIVM TTREVEKGRN KCVPYWPEVG MQRAYGPYSV TNCGEHDTTE YKLRTLQVSP LDNGDLIREI WHYQYLSWPD HGVPSEPGGV LSFLDQINQR QESLPHAGPI IVHCSAGIGR TGTIIVIDML MENISTKGLD CDIDIQKTIQ MVRAQRSGMV QTEAQYKFIY VAIAQFIETT KKKLEVLQSQ KGQESEYGNI TYPPAMKNAH AKASRTSSKH KEDVYENLHT KNKREEKVKK QRSADKEKSK GSLKRK | |
Greater than 90% as determined by SDS-PAGE. |
5777 | |
50 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |