missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTGDS / L-PGDS Protein

A DNA sequence encoding the Homo sapiens (Human) PTGDS / L-PGDS, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6550-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054090

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_000945.3
PTGDS / L-PGDS Protein
Research Use Only
PTGDS
Recombinant Protein
E. coli
APEAQVSVQP NFQQDKFLGR WFSAGLASNS SWLREKKAAL SMCKSVVAPA TDGGLNLTST FLRKNQCETR TMLLQPAGSL GSYSYRSPHW GSTYSVSVVE TDYDQYALLY SQGSKGPGED FRMATLYSRT QTPRAELKEK FTAFCKAQGF TEDTIVFLPQ TDKCMTEQ
Greater than 86% as determined by SDS-PAGE.
5730
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers