missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PSMD10 Protein
A DNA sequence encoding the Homo sapiens (Human) PSMD10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6546-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002805.1 | |
PSMD10 Protein | |
Research Use Only | |
PSMD10 | |
Recombinant Protein | |
E. coli | |
MEGCVSNLMV CNLAYSGKLE ELKESILADK SLATRTDQDS RTALHWACSA GHTEIVEFLL QLGVPVNDKD DAGWSPLHIA ASAGRDEIVK ALLGKGAQVN AVNQNGCTPL HYAASKNRHE IAVMLLEGGA NPDAKDHYEA TAMHRAAAKG NLKMIHILLY YKASTNIQDT EGNTPLHLAC DEERVEEAKL LVSQGASIYI ENKEEKTPLQ VAKGGLGLIL KRMVEG | |
Greater than 90% as determined by SDS-PAGE. |
5716 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |