missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein yippee-like 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein yippee-like 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6897-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016630

  • 2385.00€ / 1mg
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001138996.1
Protein yippee-like 3 Protein
Research Use Only
YPEL3
Recombinant Protein
E. coli
MVRISKPKTF QAYLDDCHRR YSCAHCRAHL ANHDDLISKS FQGSQGRAYL FNSVVNVGCG PAEERVLLTG LHAVADIHCE NCKTTLGWKY EQAFESSQKY KEGKYIIELN HMIKDNGWD
Greater than 90% as determined by SDS-PAGE.
83719
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers