missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein yippee-like 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein yippee-like 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6897-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001138996.1 | |
Protein yippee-like 3 Protein | |
Research Use Only | |
YPEL3 | |
Recombinant Protein | |
E. coli | |
MVRISKPKTF QAYLDDCHRR YSCAHCRAHL ANHDDLISKS FQGSQGRAYL FNSVVNVGCG PAEERVLLTG LHAVADIHCE NCKTTLGWKY EQAFESSQKY KEGKYIIELN HMIKDNGWD | |
Greater than 90% as determined by SDS-PAGE. |
83719 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |