missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein unc-119 homolog A Protein
A DNA sequence encoding the Homo sapiens (Human) Protein unc-119 homolog A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7593-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001317095.1 | |
Protein unc-119 homolog A Protein | |
Research Use Only | |
UNC119 | |
Recombinant Protein | |
E. coli | |
MKVKKGGGGA GTATESAPGP SGQSVAPIPQ PPAESESGSE SEPDAGPGPR PGPLQRKQPI GPEDVLGLQR ITGDYLCSPE ENIYKIDFVR FKIRDMDSGT VLFEIKKPPV SERLPINRRD LDPNAGRFVR YQFTPAFLRL RQVGATVEFT VGDKPVNNFR MIERHYFRNQ LLKSFDFHFG FCIPSSKNTC EHIYDFPPLS EELISEMIRH PYETQSDSFY FVDDRLVMHN KADYSYSGTP | |
Greater than 90% as determined by SDS-PAGE. |
9094 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |