missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein SSX2 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein SSX2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7535-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16019230

  • 2385.00€ / 1mg
Estimated Shipment: 01-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001157889.1
Protein SSX2 Protein
Research Use Only
SSX2
Recombinant Protein
E. coli
MNGDDAFARR PTVGAQIPEK IQKAFDDIAK YFSKEEWEKM KASEKIFYVY MKRKYEAMTK LGFKATLPPF MCNKRAEDFQ GNDLDNDPNR GNQVERPQMT FGRLQGISPK IMPKKPAEEG NDSEEVPEAS GPQNDGKELC PPGKPTTSEK IHERSGPKRG EHAWTHRLRE RKQLVIYEEI SDPEEDDE
Greater than 90% as determined by SDS-PAGE.
6757
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers